siRNA Details
| Gene Symbol: Ggct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026637.3 | NP_080913.1 | 1, 2 | 181 | ||
| GenBank mRNA | AK008719.1 | - | 1, 2 | 186 | ||
| AK028830.1 | - | 183 | ||||
| AK050049.1 | - | 1, 2 | 266 | |||
| AK079465.1 | - | 1, 2 | 183 | |||
| AK141474.1 | - | 231 | ||||
| BC080818.1 | - | 1, 2 | 161 | 30286931 | ||

Ggct
Genomic Map