siRNA Details
| Gene Symbol: Iqcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026645.3 | NP_080921.2 | 3, 4 | 232 | ||
| GenBank mRNA | AK016688.1 | - | 3, 4 | 233 | ||
| AK018835.1 | - | 3, 4 | 235 | |||
| AK132739.1 | - | 3, 4 | 237 | |||
| BC049549.1 | - | 3, 4 | 233 | 6774389 | ||
| DQ153247.1 | - | 3, 4 | 238 | |||

Iqcf3
Genomic Map