siRNA Details
| Gene Symbol: Pomgnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026651.2 | NP_080927.1 | 2, 3 | 274 | ||
| NM_029786.2 | NP_084062.2 | 2, 3 | 274 | |||
| GenBank mRNA | AB053220.1 | - | 2, 3 | 256 | ||
| AK002638.1 | - | 2, 3 | 266 | |||
| BC011201.1 | - | 2, 3 | 197 | 4190368 | ||
| BY732188.1 | - | 2, 3 | 274 | |||

Pomgnt1
Genomic Map