siRNA Details
| Gene Symbol: PCCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000282.3 | NP_000273.2 | 3 | 328 | ||
| NM_001127692.2 | NP_001121164.1 | 2 | 250 | |||
| NM_001178004.1 | NP_001171475.1 | 3 | 328 | |||
| XM_005254059.2 | XP_005254116.1 | 3 | 323 | |||
| XM_011521093.1 | XP_011519395.1 | 3 | 323 | |||
| XM_017020605.1 | XP_016876094.1 | 3 | 323 | |||
| XM_017020606.1 | XP_016876095.1 | 2 | 174 | |||
| XM_017020607.1 | XP_016876096.1 | 3 | 139 | |||
| XM_017020608.1 | XP_016876097.1 | 3 | 323 | |||
| XM_017020609.1 | XP_016876098.1 | 3 | 236 | |||
| XM_017020611.1 | XP_016876100.1 | 3 | 323 | |||
| XM_017020612.1 | XP_016876101.1 | 3 | 323 | |||
| XM_017020613.1 | XP_016876102.1 | 3 | 323 | |||
| XM_017020614.1 | XP_016876103.1 | 2 | 174 | |||
| XM_017020615.1 | XP_016876104.1 | 3 | 323 | |||
| XM_017020616.1 | XP_016876105.1 | 3 | 323 | |||
| GenBank mRNA | AF385926.1 | - | 3 | 260 | ||
| AK225288.1 | - | 3 | 228 | |||
| AK225299.1 | - | 3 | 226 | |||
| AK296771.1 | - | 2 | 148 | |||
| AK298318.1 | - | 3 | 250 | |||
| AK315503.1 | - | 3 | 226 | |||
| BC000140.2 | - | 3 | 200 | 3048998 | ||
| DA781564.1 | - | 2 | 148 | |||
| X14608.1 | - | 3 | 195 | |||

PCCA
Genomic Map