siRNA Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106.3 | NP_001097.2 | 5 | 650 | ||
| XM_005265583.3 | XP_005265640.1 | 5 | 3112 | |||
| XM_017007514.1 | XP_016863003.1 | 5 | 3091 | |||
| XM_017007515.1 | XP_016863004.1 | 5 | 736 | |||
| XM_017007516.1 | XP_016863005.1 | 5 | 4060 | |||
| GenBank mRNA | AB209449.1 | - | 4 | 4415 | ||
| BC096243.1 | - | 5 | 630 | 40005760 | ||
| BC096244.1 | - | 5 | 630 | 40005761 | ||
| BC096245.1 | - | 4 | 712 | 40005763 | ||
| BC099642.4 | - | 5 | 630 | 40005762 | ||
| X77533.1 | - | 5 | 630 | |||

ACVR2B
Genomic Map