siRNA Details
| Gene Symbol: Tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163718.1 | NP_001157190.1 | 4, 5 | 763 | ||
| NM_026676.3 | NP_080952.2 | 4, 5 | 784 | |||
| GenBank mRNA | AK002309.1 | - | 4, 5 | 756 | ||
| AK158743.1 | - | 4, 5 | 785 | |||
| AK170074.1 | - | 4, 5 | 753 | |||
| BC006899.1 | - | 1, 2 | 129 | |||
| BC027751.1 | - | 4, 5 | 854 | 5011894 | ||
| BC089019.1 | - | 4, 5 | 733 | 30551363 | ||
| CA979987.1 | - | 4, 5 | 712 | |||

Tsr3
Genomic Map