siRNA Details
| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026695.3 | NP_080971.2 | 2 | 243 | ||
| GenBank mRNA | AA207939.1 | - | 2 | 127 | ||
| AK002407.1 | - | 2 | 185 | |||
| AK013284.1 | - | 2 | 208 | |||
| AK150293.1 | - | 2 | 186 | |||
| BC049237.1 | - | 2 | 172 | 4241552 | ||
| BC069877.1 | - | 2 | 202 | 5370580 | ||

Etfb
Genomic Map