siRNA Details
| Gene Symbol: Ndufa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026703.2 | NP_080979.1 | 2 | 181 | ||
| GenBank mRNA | AK002744.1 | - | 2 | 181 | ||
| AK088463.1 | - | 2 | 227 | |||
| BC012416.1 | - | 2 | 179 | 3584066 | ||
| BC024673.1 | - | 2 | 178 | 4195454 | ||

Ndufa8
Genomic Map