siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 2, 3 | 305 | ||
| NM_026728.4 | NP_081004.2 | 3, 4 | 373 | |||
| GenBank mRNA | AK005030.1 | - | 3, 4 | 314 | ||
| AK011365.1 | - | 2, 3 | 251 | |||
| AK153878.1 | - | 3, 4 | 348 | |||
| AK166388.1 | - | 3, 4 | 345 | |||
| AK166965.1 | - | 3, 4 | 315 | |||
| BC025104.1 | - | 3, 4 | 319 | 3591648 | ||

Echdc2
Genomic Map