siRNA Details
| Gene Symbol: Mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026744.3 | NP_081020.1 | 3 | 264 | ||
| GenBank mRNA | AK003549.1 | - | 3 | 265 | ||
| BC022162.1 | - | 3 | 289 | 5148541 | ||
| BC065995.1 | - | 1487 | 5688315 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl53| Gene Symbol: Mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026744.3 | NP_081020.1 | 3 | 264 | ||
| GenBank mRNA | AK003549.1 | - | 3 | 265 | ||
| BC022162.1 | - | 3 | 289 | 5148541 | ||
| BC065995.1 | - | 1487 | 5688315 | |||