siRNA Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026609.2 | NP_080885.1 | 4 | 444 | ||
| GenBank mRNA | AK003329.1 | - | 4 | 455 | ||
| AK013870.1 | - | 4 | 514 | |||
| AK075740.1 | - | 4 | 499 | |||
| AK150289.1 | - | 4 | 503 | |||
| AK153277.1 | - | 4 | 489 | |||
| AK154712.1 | - | 4 | 509 | |||
| AK159401.1 | - | 4 | 432 | |||
| BC004677.1 | - | 4 | 444 | 3500223 | ||

Leprotl1
Genomic Map