siRNA Details
Gene Symbol: Wdr86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081441.1 | NP_001074910.1 | 6 | 1675 | ||
GenBank mRNA | AK019251.1 | - | 1 | 442 | ||
AK019267.1 | - | 1 | 442 | |||
BC059069.1 | - | 5 | 4247 | |||
BY712199.1 | - | 1 | 436 |
