siRNA Details
| Gene Symbol: 3110057O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026622.3 | NP_080898.2 | 1332 | |||
| GenBank mRNA | AK028580.1 | - | 1349 | |||
| AK031032.1 | - | 1152 | ||||
| AK163334.1 | - | 3 | 513 | |||
| AK169375.1 | - | 1333 | ||||
| BC051081.1 | - | 1402 | ||||
| BC092533.1 | - | 2 | 372 | 30503250 | ||
| BC145212.1 | - | 839 | 9053763 | |||

3110057O12Rik
Genomic Map