siRNA Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026647.3 | NP_080923.2 | 9 | 1220 | ||
| GenBank mRNA | AK020293.1 | - | 9 | 1102 | ||
| AK144108.1 | - | 8 | 982 | |||
| BC043110.1 | - | 9 | 1066 | 6406930 | ||
| BC058372.1 | - | 9 | 1087 | |||

Zdhhc21
Genomic Map