siRNA Details
| Gene Symbol: Fam114a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026667.3 | NP_080943.3 | 6 | 768 | ||
| GenBank mRNA | AK007950.1 | - | 6 | 726 | ||
| AK014725.1 | - | 6 | 738 | |||
| AK017336.1 | - | 6 | 728 | |||
| AK020262.1 | - | 6 | 729 | |||
| AK030360.1 | - | 6 | 769 | |||
| AM162548.1 | - | 6 | 772 | |||
| BC027235.1 | - | 6 | 731 | 3603083 | ||
| BC110359.1 | - | 6 | 769 | 5325099 | ||

Fam114a1
Genomic Map