siRNA Details
| Gene Symbol: Lrriq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290510.1 | NP_001277439.1 | 2 | 606 | ||
| NM_026668.2 | NP_080944.2 | 4 | 769 | |||
| GenBank mRNA | AK015024.1 | - | 2 | 557 | ||
| AK019775.1 | - | 4 | 770 | |||
| BC145806.1 | - | 4 | 470 | 40131109 | ||

Lrriq4
Genomic Map