siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 6 | 605 | ||
| NM_026728.4 | NP_081004.2 | 7 | 673 | |||
| GenBank mRNA | AK005030.1 | - | 7 | 614 | ||
| AK011365.1 | - | 6 | 551 | |||
| AK153878.1 | - | 7 | 648 | |||
| AK160482.1 | - | 5 | 376 | |||
| AK166388.1 | - | 7 | 645 | |||
| AK166965.1 | - | 7 | 615 | |||
| BC025104.1 | - | 7 | 619 | 3591648 | ||
| BC025454.1 | - | 3 | 157 | 5352625 | ||

Echdc2
Genomic Map