siRNA Details
| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164604.1 | NP_001158076.1 | 3 | 420 | ||
| NM_026746.3 | NP_081022.2 | 3 | 420 | |||
| GenBank mRNA | AK003685.1 | - | 3 | 372 | ||
| AK051277.1 | - | 3 | 372 | |||
| AK188685.1 | - | 3 | 301 | |||
| AK211545.1 | - | 3 | 301 | |||
| AK212165.1 | - | 3 | 301 | |||
| BC009125.1 | - | 3 | 331 | 3155917 | ||
| DV057668.1 | - | 3 | 440 | |||

Nsmce2
Genomic Map