siRNA Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026784.3 | NP_081060.2 | 4 | 592 | ||
| NM_027348.1 | NP_081624.1 | 4 | 561 | |||
| GenBank mRNA | AK003607.1 | - | 4 | 395 | ||
| AK013477.1 | - | 4 | 561 | |||
| AK150483.1 | - | 4 | 511 | |||
| AK168358.1 | - | 4 | 487 | |||
| BC028659.1 | - | 3 | 380 | 1247653 | ||

Pmvk
Genomic Map