siRNA Details
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177576.1 | NP_001171047.1 | 9 | 1001 | ||
| NM_026646.3 | NP_080922.1 | 9 | 1243 | |||
| GenBank mRNA | AK010193.1 | - | 9 | 989 | ||
| AK018760.1 | - | 9 | 1026 | |||
| AK046874.1 | - | 9 | 988 | |||
| AK048389.1 | - | 9 | 972 | |||
| AK049313.1 | - | 9 | 972 | |||
| AK155276.1 | - | 9 | 995 | |||
| AK158985.1 | - | 1 | 3958 | |||
| AK162400.1 | - | 9 | 954 | |||
| AK181898.1 | - | 3 | 319 | |||
| BC037949.1 | - | 9 | 1000 | 5367108 | ||
| BC050887.1 | - | 8 | 1719 | |||

Slc25a22
Genomic Map