siRNA Details
| Gene Symbol: Tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163718.1 | NP_001157190.1 | 6 | 1016 | ||
| NM_026676.3 | NP_080952.2 | 6 | 1037 | |||
| GenBank mRNA | AK002309.1 | - | 6 | 1009 | ||
| AK003675.1 | - | 3 | 1785 | |||
| AK158743.1 | - | 6 | 1038 | |||
| AK169072.1 | - | 5 | 1372 | |||
| AK170074.1 | - | 6 | 1006 | |||
| BC006899.1 | - | 3 | 382 | |||
| BC027751.1 | - | 6 | 1107 | 5011894 | ||
| BC038884.1 | - | 3 | 729 | 1516275 | ||
| BC089019.1 | - | 6 | 986 | 30551363 | ||

Tsr3
Genomic Map