siRNA Details
| Gene Symbol: Tspan11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026743.3 | NP_081019.1 | 8 | 1732 | ||
| GenBank mRNA | AK028808.1 | - | 8 | 1623 | ||
| AK053788.1 | - | 8 | 1719 | |||
| AK090022.1 | - | 8 | 1733 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan11| Gene Symbol: Tspan11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026743.3 | NP_081019.1 | 8 | 1732 | ||
| GenBank mRNA | AK028808.1 | - | 8 | 1623 | ||
| AK053788.1 | - | 8 | 1719 | |||
| AK090022.1 | - | 8 | 1733 | |||