siRNA Details
| Gene Symbol: Ucma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113558.2 | NP_001107030.1 | 5 | 847 | ||
| NM_001165932.1 | NP_001159404.1 | 4 | 748 | |||
| NM_026754.3 | NP_081030.1 | 4 | 781 | |||
| GenBank mRNA | AK003750.1 | - | 4 | 723 | ||
| BY590633.1 | - | 1 | 423 | |||
| EU368184.1 | - | 5 | 847 | |||

Ucma
Genomic Map