siRNA Details
Gene Symbol: 0610010K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177601.1 | NP_001171072.1 | 5 | 584 | ||
NM_026757.2 | NP_081033.1 | 4 | 482 | |||
NM_145758.1 | NP_665701.1 | 5 | 584 | |||
GenBank mRNA | AA189586.1 | - | 2 | 90 | ||
AK003842.1 | - | 4 | 482 | |||
AK178853.1 | - | 302 | ||||
AK208696.1 | - | 312 | ||||
CK791552.1 | - | 5 | 563 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001127521, XM_006246652, XM_006246655, XM_006246653, BC158879, and others.
Panther Classification:
Gene Ontology Categories:
