siRNA Details
| Gene Symbol: Mrps18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026768.3 | NP_081044.1 | 5 | 434 | ||
| GenBank mRNA | AB049953.1 | - | 5 | 443 | ||
| AK003444.1 | - | 5 | 435 | |||
| AK028226.1 | - | 5 | 436 | |||
| AK172171.1 | - | 5 | 429 | |||
| BC025456.1 | - | 3 | 230 | |||
| BC041151.1 | - | 2 | 183 | |||
| BC058959.1 | - | 5 | 422 | 6817698 | ||

Mrps18a
Genomic Map