siRNA Details
| Gene Symbol: Dhrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026819.3 | NP_081095.2 | 2 | 364 | ||
| GenBank mRNA | AK003958.1 | - | 2 | 254 | ||
| AK146317.1 | - | 2 | 311 | |||
| AK159964.1 | - | 2 | 286 | |||
| AK168115.1 | - | 2 | 311 | |||
| AK168147.1 | - | 2 | 311 | |||
| BC003930.1 | - | 2 | 295 | 3485584 | ||
| BC023825.1 | - | 2 | 290 | |||
| BU963142.1 | - | 367 | ||||

Dhrs1
Genomic Map