siRNA Details
| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026825.3 | NP_081101.3 | 22 | 1881 | ||
| GenBank mRNA | AK043591.1 | - | 7 | 461 | ||
| AK049059.1 | - | 4 | 257 | |||
| AY437876.1 | - | 22 | 1886 | |||
| BC150819.1 | - | 21 | 1795 | |||

Lrrc16a
Genomic Map