siRNA Details
| Gene Symbol: Ptges3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026865.2 | NP_081141.1 | 2 | 807 | ||
| GenBank mRNA | AK004415.1 | - | 2 | 125 | ||
| AK007198.1 | - | 2 | 806 | |||
| BC057970.1 | - | 2 | 113 | |||
| BC141198.1 | - | 2 | 226 | 9055810 | ||

Ptges3l
Genomic Map