siRNA Details
| Gene Symbol: Disp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278218.1 | NP_001265147.1 | 6 | 1054 | ||
| NM_001278219.1 | NP_001265148.1 | 6 | 1014 | |||
| NM_001278220.1 | NP_001265149.1 | 5 | 897 | |||
| NM_026866.3 | NP_081142.3 | 7 | 1142 | |||
| GenBank mRNA | AK004521.1 | - | 5 | 872 | ||
| AK138276.1 | - | 7 | 1099 | |||
| AK170113.1 | - | 6 | 1009 | |||
| AY144589.1 | - | 5 | 852 | |||
| AY150577.1 | - | 5 | 801 | |||
| AY150698.1 | - | 7 | 1034 | |||
| BC043102.1 | - | 6 | 1015 | 6406376 | ||
| BC059225.1 | - | 6 | 1015 | 6406376 | ||

Disp1
Genomic Map