siRNA Details
| Gene Symbol: Haghl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271433.1 | NP_001258362.1 | 4 | 516 | ||
| NM_001271434.1 | NP_001258363.1 | 4 | 516 | |||
| NM_001271435.1 | NP_001258364.1 | 4 | 516 | |||
| NM_001271436.1 | NP_001258365.1 | 3 | 330 | |||
| NM_001271437.1 | NP_001258366.1 | 3 | 325 | |||
| NM_026897.3 | NP_081173.1 | 3 | 501 | |||
| GenBank mRNA | AK005274.1 | - | 3 | 497 | ||
| AK012748.1 | - | 4 | 513 | |||
| AK021220.1 | - | 3 | 327 | |||
| AK032999.1 | - | 3 | 307 | |||
| AK034798.1 | - | 4 | 491 | |||
| AK047517.1 | - | 4 | 471 | |||
| AK079632.1 | - | 3 | 499 | |||
| AK080454.1 | - | 1 | 1097 | |||
| AK154248.1 | - | 3 | 306 | |||
| BC005586.1 | - | 4 | 462 | |||
| BC030466.1 | - | 4 | 501 | 5253213 | ||
| BC083322.1 | - | 4 | 455 | 6820885 | ||

Haghl
Genomic Map