siRNA Details
| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026907.3 | NP_081183.3 | 4 | 489 | ||
| GenBank mRNA | AF210700.1 | - | 3 | 392 | ||
| AK007321.1 | - | 3 | 351 | |||
| AK029082.1 | - | 4 | 490 | |||
| AK143807.1 | - | 4 | 473 | |||
| BC010805.1 | - | 4 | 566 | 4239879 | ||
| BC020159.1 | - | 3 | 440 | 4016267 | ||

Sectm1b
Genomic Map