siRNA Details
| Gene Symbol: Rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026967.4 | NP_081243.1 | 2 | 321 | ||
| GenBank mRNA | AK007708.1 | - | 2 | 171 | ||
| AK135858.1 | - | 2 | 276 | |||
| AY327413.1 | - | 2 | 369 | |||
| BC016521.1 | - | 2 | 240 | 3487009 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_182825, XM_008765754, XM_008765755, AY327411.

Rhebl1
Genomic Map