siRNA Details
| Gene Symbol: Zfp688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026999.4 | NP_081275.3 | 2, 3 | 348 | ||
| GenBank mRNA | AK013037.1 | - | 1, 2 | 628 | ||
| BC027683.1 | - | 1, 2 | 631 | 1345974 | ||
| BQ550868.1 | - | 2, 3 | 257 | |||
| DV654233.1 | - | 2, 3 | 349 | |||

Zfp688
Genomic Map