siRNA Details
| Gene Symbol: Tescl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163810.1 | NP_001157282.1 | 1 | 150 | ||
| GenBank mRNA | AK005787.1 | - | 1 | 124 | ||
| BC125456.1 | - | 1 | 111 | |||
| BC125458.1 | - | 1 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tescl| Gene Symbol: Tescl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163810.1 | NP_001157282.1 | 1 | 150 | ||
| GenBank mRNA | AK005787.1 | - | 1 | 124 | ||
| BC125456.1 | - | 1 | 111 | |||
| BC125458.1 | - | 1 | 111 | |||