siRNA Details
| Gene Symbol: Tmem216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277860.1 | NP_001264789.1 | 5 | 563 | ||
| NM_001277861.1 | NP_001264790.1 | 5 | 577 | |||
| NM_026798.3 | NP_081074.1 | 5 | 636 | |||
| GenBank mRNA | AF332866.1 | - | 4 | 502 | ||
| AK003740.1 | - | 5 | 638 | |||
| AK013286.1 | - | 5 | 567 | |||
| BC052053.1 | - | 5 | 540 | 5693205 | ||
| BY704000.1 | - | 5 | 638 | |||
| DV063021.1 | - | 5 | 631 | |||

Tmem216
Genomic Map