siRNA Details
| Gene Symbol: Krtap4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026807.2 | NP_081083.2 | 1 | 826 | ||
| GenBank mRNA | AK004055.1 | - | 1 | 827 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap4-2| Gene Symbol: Krtap4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026807.2 | NP_081083.2 | 1 | 826 | ||
| GenBank mRNA | AK004055.1 | - | 1 | 827 | ||