siRNA Details
| Gene Symbol: Hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026812.2 | NP_081088.1 | 3 | 228 | ||
| GenBank mRNA | AK004078.1 | - | 3 | 229 | ||
| AK080642.1 | - | 27 | ||||
| BC038310.1 | - | 3 | 241 | 5119020 | ||
| BC055847.1 | - | 2 | 530 | |||

Hddc3
Genomic Map