siRNA Details
| Gene Symbol: Cd177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026862.3 | NP_081138.2 | 16 | 2181 | ||
| GenBank mRNA | AK004472.1 | - | 3 | 341 | ||
| BC005615.1 | - | 11 | 1551 | |||
| BC027283.1 | - | 16 | 2181 | 2648499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd177| Gene Symbol: Cd177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026862.3 | NP_081138.2 | 16 | 2181 | ||
| GenBank mRNA | AK004472.1 | - | 3 | 341 | ||
| BC005615.1 | - | 11 | 1551 | |||
| BC027283.1 | - | 16 | 2181 | 2648499 | ||