siRNA Details
| Gene Symbol: Tmco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039483.1 | NP_001034572.1 | 2 | 322 | ||
| GenBank mRNA | AK004504.1 | - | 2 | 164 | ||
| AK015068.1 | - | 2 | 613 | |||
| AK037709.1 | - | 2 | 257 | |||
| AK162344.1 | - | 2 | 261 | |||
| AK183030.1 | - | 2 | 75 | |||
| AK218449.1 | - | 2 | 75 | |||
| AK219964.1 | - | 2 | 75 | |||
| BC011457.1 | - | 2 | 196 | 3585288 | ||
| BC019761.1 | - | 2 | 240 | 5009960 | ||
| BC020098.1 | - | 2 | 252 | 4019484 | ||

Tmco1
Genomic Map