siRNA Details
| Gene Symbol: Chmp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026885.3 | NP_081161.1 | 2 | 314 | ||
| GenBank mRNA | AK005267.1 | - | 2 | 282 | ||
| BC012230.1 | - | 2 | 255 | 4024872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp2a| Gene Symbol: Chmp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026885.3 | NP_081161.1 | 2 | 314 | ||
| GenBank mRNA | AK005267.1 | - | 2 | 282 | ||
| BC012230.1 | - | 2 | 255 | 4024872 | ||