siRNA Details
| Gene Symbol: Ssu72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026899.3 | NP_081175.2 | 2, 3 | 784 | ||
| GenBank mRNA | AK005220.1 | - | 2, 3 | 475 | ||
| AK019168.1 | - | 2, 3 | 240 | |||
| AK148783.1 | - | 1, 2 | 81 | |||
| BC016544.1 | - | 2, 3 | 464 | 3497878 | ||

Ssu72
Genomic Map