siRNA Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026935.4 | NP_081211.3 | 2, 3 | 368 | ||
| GenBank mRNA | AK007384.1 | - | 2, 3 | 228 | ||
| AK144336.1 | - | 2, 3 | 308 | |||
| AY005469.1 | - | 2, 3 | 368 | |||
| BC022665.1 | - | 2, 3 | 361 | 4223678 | ||

Sult1c2
Genomic Map