siRNA Details
| Gene Symbol: Kbtbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164574.1 | NP_001158046.1 | 3 | 524 | ||
| NM_026962.3 | NP_081238.2 | 4 | 556 | |||
| GenBank mRNA | AK008627.1 | - | 3 | 414 | ||
| AK034334.1 | - | 4 | 464 | |||
| AK078744.1 | - | 3 | 436 | |||
| AK083497.1 | - | 4 | 538 | |||
| AK168345.1 | - | 3 | 370 | |||
| BB264869.2 | - | 3 | 525 | |||
| BC024938.1 | - | 3 | 420 | |||
| BC027392.1 | - | 4 | 460 | 4954712 | ||
| BE650264.1 | - | 4 | 557 | |||

Kbtbd3
Genomic Map