siRNA Details
| Gene Symbol: Zfp688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026999.4 | NP_081275.3 | 3 | 434 | ||
| GenBank mRNA | AK013037.1 | - | 2 | 714 | ||
| AK170857.1 | - | 2 | 2156 | |||
| BC027683.1 | - | 2 | 717 | 1345974 | ||
| BC052512.1 | - | 3 | 274 | 1362502 | ||
| BQ550868.1 | - | 3 | 343 | |||
| DV654233.1 | - | 3 | 436 | |||

Zfp688
Genomic Map