siRNA Details
| Gene Symbol: Hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026812.2 | NP_081088.1 | 2 | 147 | ||
| GenBank mRNA | AK004078.1 | - | 2 | 148 | ||
| BC038310.1 | - | 2 | 160 | 5119020 | ||
| BC055847.1 | - | 1 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hddc3| Gene Symbol: Hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026812.2 | NP_081088.1 | 2 | 147 | ||
| GenBank mRNA | AK004078.1 | - | 2 | 148 | ||
| BC038310.1 | - | 2 | 160 | 5119020 | ||
| BC055847.1 | - | 1 | 449 | |||