siRNA Details
Gene Symbol: Fam216a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026883.3 | NP_081159.1 | 5, 6 | 571 | ||
GenBank mRNA | AK005211.1 | - | 5, 6 | 963 | ||
AK182306.1 | - | 1, 2 | 99 | |||
BC019498.1 | - | 5, 6 | 929 | 4460879 | ||
BC057658.1 | - | 5, 6 | 610 | 3153717 | ||
BC061033.1 | - | 5, 6 | 571 | 6590939 |
