siRNA Details
| Gene Symbol: Nupr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026916.3 | NP_081192.2 | 3 | 519 | ||
| GenBank mRNA | AK007417.1 | - | 3 | 494 | ||
| AK016320.1 | - | 3 | 514 | |||
| BC100444.1 | - | 3 | 519 | |||
| EL963130.1 | - | 3 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nupr1l| Gene Symbol: Nupr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026916.3 | NP_081192.2 | 3 | 519 | ||
| GenBank mRNA | AK007417.1 | - | 3 | 494 | ||
| AK016320.1 | - | 3 | 514 | |||
| BC100444.1 | - | 3 | 519 | |||
| EL963130.1 | - | 3 | 412 | |||