siRNA Details
| Gene Symbol: 1810011O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026931.2 | NP_081207.1 | 1 | 836 | ||
| GenBank mRNA | AK007441.1 | - | 1 | 837 | ||
| BC016562.1 | - | 1 | 793 | 3154083 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810011O10Rik| Gene Symbol: 1810011O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026931.2 | NP_081207.1 | 1 | 836 | ||
| GenBank mRNA | AK007441.1 | - | 1 | 837 | ||
| BC016562.1 | - | 1 | 793 | 3154083 | ||