siRNA Details
| Gene Symbol: Fastkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027123.4 | NP_081399.3 | 2 | 341 | ||
| GenBank mRNA | AK031142.1 | - | 2 | 328 | ||
| AK032530.1 | - | 2 | 326 | |||
| AK033189.1 | - | 2 | 342 | |||
| AK036778.1 | - | 2 | 327 | |||
| AK148902.1 | - | 2 | 331 | |||
| BC042506.1 | - | 2 | 318 | 4023412 | ||
| BC060688.1 | - | 2 | 561 | 6409274 | ||

Fastkd3
Genomic Map