siRNA Details
Gene Symbol: Cdk3-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009918.1 | - | 4 | 582 | ||
AK166387.1 | - | 1 | 69 | |||
BC116895.1 | - | 4 | 492 | |||
BC119034.1 | - | 4 | 492 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdk3-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009918.1 | - | 4 | 582 | ||
AK166387.1 | - | 1 | 69 | |||
BC116895.1 | - | 4 | 492 | |||
BC119034.1 | - | 4 | 492 |